missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SILBERBRAND Automatic Burette Spare Burette Tube

Product Code. 10606331 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Color:
Amber
Clear
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Unit Size
10606331 25 mL Clear Each
10552964 25 mL Amber Each
10218464 50 mL Amber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10606331 Supplier BRAND™ Supplier No. 23716
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

SILBERBRAND Spare Burette Tube is suitable for use with an automatic burette.

  • For use with automatic burette
  • Dr. Scilling pattern
TRUSTED_SUSTAINABILITY

Specifications

Description Spare burette f. aut.burette Schilling 25 ml, SILBERBRAND, Schellbach, Borosilicate Glass 3.3, length 380 mm
Color Clear
Capacity (Metric) 25 mL
Length (Metric) 380 mm
For Use With (Equipment) Automatic Burette
Material Borosilicate Glass (3.3)
Type Spare Buret
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.