missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SILBERBRAND Automatic Burette Spare Burette Tube

Product Code. p-7164794 Shop All Brand Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
Color:
Amber
Clear
Conditionnement:
Each
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 10606331

Marque: BRAND™ 23716

Il en reste null
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

SILBERBRAND Spare Burette Tube is suitable for use with an automatic burette.

  • For use with automatic burette
  • Dr. Scilling pattern
TRUSTED_SUSTAINABILITY

Spécification

Description Spare burette f. aut.burette Schilling 25 ml, SILBERBRAND, Schellbach, Borosilicate Glass 3.3, length 380 mm
Color Clear
Capacity (Metric) 25 mL
Length (Metric) 380 mm
For Use With (Equipment) Automatic Burette
Material Borosilicate Glass (3.3)
Type Spare Buret
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.