Learn More
BRAND™ SilberBrand™ Burette Tube for Compact Burette
Product Code. 10404732 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
10404732 50 mL Each
1 options
Product Code. 10404732 Supplier BRAND™ Supplier No. 13938
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Borosilicate glass construction. Replacement burette tube for compact burette.
  • SilberBrand™ borosilicate glass (3.3) construction
  • Replacement burette tube
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
For Use With (Equipment) Compact Burette
Material Borosilicate Glass
Type Buret Tube