missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ SilberBrand™ Compact-Burette (Boro 5.4)
With PTFE stopcock, fast to dismantle and easy to clean
93.20 €
Specifications
Color | Amber |
---|---|
Material | Borosilicate Glass |
Graduations | 0.1 mL |
Stopcock Material | PTFE |
Type | Compact Buret |
Description
Specifications
Amber | |
0.1 mL | |
Compact Buret | |
DIN EN ISO 385 |
Borosilicate Glass | |
PTFE | |
Class B |