missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Negative Control No. 1 siRNA

Product Code. 10670226
Change view
Click to view available options
Quantity:
250 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10670226 250 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10670226 Supplier Invitrogen™ Supplier No. 4404021
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Ambion™ Silencer Negative Control No. 1 siRNA, In Vivo Ready has no significant sequence similarity to mouse, rat, or human gene sequences.

Ambion™ Silencer Negative Control No. 1 siRNA, In Vivo Ready has no significant sequence similarity to mouse, rat, or human gene sequences. The control has also been tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology. It is highly purified as required for animal use. 250 nmol is provided.

  • Nontargeting with limited sequence similarity to known genes
  • Validated for use in human, mouse, and rat cells
  • Functionally proven to have minimal effects on cell proliferation and viability
  • HPLC purified, duplexed, and ready to use

Negative control siRNAs are essential for determining transfection efficiency and to control for the effects of siRNA delivery. In siRNA screening applications, negative control siRNAs are also critical for setting the 'hit' threshold that determines whether an siRNA is considered to have a positive, negative, or neutral effect in a particular assay. Silencer™ Negative Control No. 1 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences. The control has also been tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology.

Quality Control:

Silencer™ siRNAs are manufactured in a state-of-the-art facility under the highest quality standards. After HPLC purification and annealing, each siRNA is further purified utilizing a process that removes excess salt via a semipermeable membrane. The result is highly pure siRNA with minimal salt content, suitable for in vivo applications. In Vivo Ready siRNAs are then sterile filtered and tested for the presence of endotoxin. At concentrations of 50 μM in deionized water, In Vivo Ready siRNAs contain less than 0.6 mM Na+, less than 2.0 mM K+, and less than 0.1 mM Mg2+.

Accessory Product: Silencer™ siRNA Screening Control Panel (Cat. No. AM4640)

The Silencer™ siRNA Screening Control Panel includes seven negative control siRNAs, and is for those researchers performing siRNA screens that employ hundreds or thousands of siRNAs per experiment. Because negative control siRNAs may have unintended effects in a particular assay, a negative control siRNA should be carefully chosen. The Silencer™ siRNA Screening Control Panel simplifies this process. As an added benefit, a positive control siRNA to human, mouse, and rat KIF11 (Eg5) is included in the panel. Knockdown of KIF11, which encodes a kinesin family motor protein, leads to mitotic arrest. Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Ambion™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The siRNA is provided dried down in a single tube, which should be stored at -20°C.
Control Type Negative Control
RNAi Type siRNA
For Use With (Application) Epigenetics and Non-Coding RNA Research, RNAi, siRNA
Label or Dye Unconjugated
Product Line Silencer, Ambion
Product Type siRNA
Purity HPLC
Quantity 250 nmol
Shipping Condition Room Temperature
How critical is it to include positive and negative controls in siRNA experiments?

A positive control siRNA such as the Silencer Select GAPDH siRNA will demonstrate efficient transfection and help to optimize transfection conditions. A negative control siRNA is necessary to serve as a baseline for gene expression.

What are the positive and negative controls I should use with my Silencer or Silencer Select siRNA experiment?

We offer several negative and positive controls for both Silencer and Silencer Select siRNA.

For Silencer siRNA positive control:
- Silencer Firefly Luciferase (GL2 + GL3) siRNA (Cat. No. AM4629, 5nmol)
- Silencer GAPDH siRNA (human) (Cat. No AM4605, 5nmol; AM4633, 40 nmol)
- Silencer GAPDH siRNA (human, mouse, rat) (Cat. No. AM4624, 5 nmol; AM4631, 40 nmol; AM4632, 5 x 40 nmol)
- Silencer GFP (eGFP) siRNA (Cat. No. AM4626, 5 nmol)
- Silencer KIF11 (Eg5) siRNA (human, mouse, rat) (Cat. No. AM4639, 5 nmol)

If working with Fluorescently labeled Silencer siRNA:
- Silencer Cy-3 labeled GAPDH siRNA (human, mouse, rat) (Cat. No. AM4649, 5 nmol)
- Silencer FAM-labeled GAPDH siRNA (human, mouse, rat) (Cat. NO. AM4650, 5 nmol)

If working with Silencer siRNA in vivo ready:
- Silencer GAPDH Positive Control siRNA, in vivo ready (Cat. No. 4404025, 250 nmol)

For Silencer siRNA negative controls:
- Silencer Negative Control No. 1 siRNA (Cat. Nos. AM4611, 5 nmol; AM4635, 40 nmol; AM4636, 5 x 40 nmol)
- Silencer Negative Control No. 2 siRNA (Cat. Nos. AM4613, 5 nmol; AM4637, 30 nmol)
- Silencer Negative Control No. 3 siRNA (Cat. No. AM4615, 5 nmol)
- Silencer Negative Control No. 4 siRNA (Cat. No. AM4641, 5 nmol)
- Silencer Negative Control No. 5 siRNA (Cat. No. AM4642, 5 nmol)

If working with Fluorescently labeled Silencer siRNA:
- Silencer Cy 3-labeled Negative Control No. 1 siRNA (Cat. No AM4621, 5 nmol)
- Silencer FAM-labeled Negative Control No. 1 siRNA (Cat. No. AM4620, 5 nmol)

If working with Silencer siRNA in vivo ready:
- Silencer in vivo ready (Cat. No. 4404021)

For Silencer Select siRNA Positive control:
- Silencer Select GAPDH Positive Control siRNA (Cat. No. 4390849, 5 nmol; 4390850, 40 nmol; 4404024, 250 nmol)
- Silencer Select MALAT1 Positive Control siRNA for non-coding siRNA (Cat. No. 4455877, 5 nmol)

For Silencer Select siRNA negative control:
- Silencer Select Negative Control No. 1 siRNA (Cat. No. 4390843, 5 nmol; 4390844, 40 nmol)
- Silencer Select Negative Control No. 2 siRNA (Cat. No. 4390846, 5 nmol; 4390847, 40 nmol)

If working with Silencer Select siRNA in vivo ready:
- Silencer Select Negative Control No. 1 siRNA, in vivo ready (Cat. No. 4404020, 250nmol)

What is the nucleotide sequence of the Silencer Negative Control No. 1 siRNA (Cat. No. 4404021)?

The sequence of Silencer Negative Control No. 1 siRNA or any other control siRNA is proprietary.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.