missing translation for 'onlineSavingsMsg'
Learn More

Silica gel 60, with fluorescent indicator, 0.060-0.2mm (70-230 mesh)

Product Code. 11375557
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Unit Size:
1kg
250g
50g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375557 50 g 50g
11355557 250 g 250g
11365557 1 kg 1kg
11385557 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11375557 Supplier Thermo Scientific Alfa Aesar Supplier No. 042757.18
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

Dry column chromatography without applied pressure (gravity pulled) | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica gel 60 is used as carriers, processing aids, anti-caking and free-flow agents in animal feed. Defoamer applications such as paint, food, paper, textile and other industrial applications. Synthetic silicon dioxides are used as a rheology control agent in plastics. It is also used in dry column chromatography without applied pressure (gravity pulled).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form -70+230 Mesh Powder,S.A. 500-600m
Material Silica Gel
Quantity 50 g
Mesh Size 70-230 Mesh
Odor Odorless
Particle Size 0.060 to 0.2mm
Solubility Information Insoluble in water.
Surface Area 500 to 600m2/g
Chemical Name or Material Silica gel 60, With fluorescent indicator

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.