missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, C18-RP, 17%C, approx. 0.7 mmol/g, part. size: 48-65 micron

Product Code. 10111002 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10111002 50 g Glass Bottle 50g
11924121 250 g Glass Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10111002 Supplier Thermo Scientific Acros Supplier No. 360120500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71889-02-6 | C18H37Cl3O2Si2 | 448.012 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71889-02-6
Molecular Formula C18H37Cl3O2Si2
Molecular Weight (g/mol) 448.012
InChI Key VYTRPUAMVCWPLO-UHFFFAOYSA-N
Synonym Octadecylsilicate
PubChem CID 172932
IUPAC Name dioxosilane;trichloro(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O

Specifications

Quantity 50 g
Packaging Glass Bottle
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Silica gel, C18-RP, 17%C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.