missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel, Solstice

Product Code. 15641360
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 500 g
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
500g
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15641360 1 kg 1kg
15661360 500 g 500g
15651360 2.5 kg 2.5kg
15691360 6 x 500 g Pack of 6
15671360 6 x 1 kg Pack of 6
15681360 6 x 2.5 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15641360 Supplier Honeywell-Fluka Supplier No. 137671KG
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With moisture indicator free of heavy metals

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112926-00-8
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Material Silica Gel
Quantity 1 kg
UN Number NONH for all modes of transport
Chemical Name or Material Silica Gel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.