missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Rubin Drying Bags, Honeywell™ Fluka™

Product Code. 15635490 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15635490 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15635490 Supplier Honeywell-Fluka Supplier No. 424061KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With indicator function, 2 TE, bag with ∼60 g, crystalline

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 2230°C
Color Red
Melting Point >1600°C
Physical Form Granular
pH ≥9
CAS 112926-00-8
Density ∽2g/cu.cm at 20°C
Material Silica Gel
Quantity 1 kg
Molecular Formula O2Si
UN Number NONH for all modes of transport
Odor Odorless
Packaging Poly Bottle
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Chemical Name or Material Silica gel Rubin drying bags
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.