missing translation for 'onlineSavingsMsg'
Learn More

Silica Gel Rubin Drying Bags, Solstice

Product Code. 15696910 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Metal Can
Paper Poly Bag
Poly Bottle
Unit Size:
1kg
250g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15696910 250 g Paper Poly Bag 250g
15667160 250 g Metal Can 250g
15657160 1 kg Metal Can 1kg
15686910 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15696910 Supplier Honeywell-Fluka Supplier No. 72811250G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With indicator function, 1/3 TE, bag with ∼10 g, crystalline

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silica gel Rubin drying bags
Boiling Point 2230°C
Color Red
Melting Point 1600°C
pH 9
CAS 112926-00-8
Density 2 g/cm3
Material Silica Gel
Quantity 250 g
UN Number NONH for all modes of transport
Packaging Paper Poly Bag
Solubility Information Slightly soluble in water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.