missing translation for 'onlineSavingsMsg'
Learn More

Silica gel, spherical, 300 angstroms

Product Code. 11359568
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359568 50 g 50g
11349568 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359568 Supplier Thermo Scientific Alfa Aesar Supplier No. 044101.18
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-67-4
Molecular Formula H2O3Si
Molecular Weight (g/mol) 78.098
MDL Number MFCD00011232
InChI Key IJKVHSBPTUYDLN-UHFFFAOYSA-N
Synonym metasilicic acid, silicic acid h2sio3, mikronisil, acidum silicicum, bio-sil, kieselsaure german, cubosic, dihydroxysilanone, silica acid, polysilicic acid
PubChem CID 14768
IUPAC Name dihydroxy(oxo)silane
SMILES O[Si](=O)O

Specifications

Quantity 50 g
Packaging Plastic bottle
Chemical Name or Material Silica gel

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.