missing translation for 'onlineSavingsMsg'
Learn More

Silica PR Tablets, Lovibond™

Product Code. 10737313
Click to view available options
Quantity:
100 Tablets
Unit Size:
100 tablets
This item is not returnable. View return policy

Product Code. 10737313

Brand: Lovibond Tintometer 513150BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Format Tablets
Quantity 100 Tablets
Type Silica PR Tablets
compliance-icons
Product Identifier
  • Silica PR
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P313-Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.