missing translation for 'onlineSavingsMsg'
Learn More

Silicon, 99+%, powder, -325 mesh

Product Code. 10092760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10092760 10 g Glass Bottle 10g
10626412 100 g Plastic bottle 100g
10732125 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092760 Supplier Thermo Scientific Acros Supplier No. 201560100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-21-3 | Si | 28.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Boiling Point 2355.0°C
Melting Point 1410.0°C
Color Gray
Physical Form Fine Powder
Quantity 10 g
Assay Percent Range 99+%
Merck Index 15, 8630
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in molten alklai oxides, soluble in a mixture of nitric acid and hydrofluor, alkalis, insoluble in nitric acid and hydrochloric acid
Formula Weight 28.09
Percent Purity ≥99%
Packaging Glass Bottle
Chemical Name or Material Silicon, -325 mesh
Show More Show Less
compliance-icons
Product Identifier
  • Silicon powder
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.