missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide, beta-phase, nanopowder, 95% min

Product Code. 11354918
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11354918 25 g 25g
11344918 5 g 5g
11334918 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354918 Supplier Thermo Scientific Alfa Aesar Supplier No. 043884.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 409-21-2 | CSi | 40.10 g/mol

Silicon carbide is used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Used in electronic applications, measure gas temperatures in an optical technique called thin filament pyrometry. Used in jewelry, steel production, grapheme production. It is also used in production of exponential resistors, wire sawing, construction industry, a filler in plastics and composites, ceramic sintered parts, ferrites and soft metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Physical Form Powder
Quantity 25 g
Odor Odorless
Merck Index 14,8492
Solubility Information Insoluble in water, alcohol; Soluble in fused alkali
IUPAC Name methanidylidynesilylium
Formula Weight 40.1
Percent Purity ≥95%
Chemical Name or Material Silicon carbide, beta-phase
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.