missing translation for 'onlineSavingsMsg'
Learn More

Silicon dioxide, 99.998%, (trace metal basis)

Product Code. 11934121 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
20 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
20g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11934121 20 g Glass bottle 20g
11944121 5 g Glass bottle 5g
11954121 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11934121 Supplier Thermo Scientific Acros Supplier No. 437150200
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For ultra trace analysis | CAS: 14808-60-7 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Melting Point 1610.0°C
Color Colorless to Yellow
Physical Form Solution
Quantity 20 g
Packaging Glass bottle
Assay Percent Range 44.5 to 47.9% (Si)
Linear Formula SiO2
Solubility Information Solubility in water: insoluble
Formula Weight 60.08
Percent Purity 99.998%
Grade Trace Metal Basis
Chemical Name or Material Silicon dioxide
Show More Show Less
compliance-icons
Product Identifier
  • Silicon dioxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.