missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chiralplate™ for TLC Enantiomer Separation
Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
5 x 20 cm
10 x 10 cm
10 x 20 cm
Quantity:
25
25 Pk.
4
4 Pk.
50
Unit Size:
Pack of 25
Pack of 4
Pack of 50
Description
Features
- Reversed phase nano silica impregnated with Cu(II) ions and a chiral selector (a proline derivative, DP 31 43 726 and EP 0 143 147)
- Separation based on ligand exchange, i. e. formation of ternary mixed-ligand complexes with the Cu(II) ions; differences in the stability of the diastereomeric complexes cause chromatographic separation
- Available as glass plates with fluorescent indicator
Application
- Enantiomer separation of amino acids, N-methylamino acids, N-formylamino acids, α-alkylamino acids, thiazolidine derivatives, dipeptides, lactones, α-hydroxycarboxylic acids
- A review on the application of CHIRALPLATE has been given by K. Günther [J. Chromatogr. 448 (1988) 11–30].
Specifications
Specifications
| Backing Material | Silica |
| Material | Silica |
| Quantity | 25 |
| Type | Glass Plate |
| Fluorescent Indicator | UV254 |
| For Use With (Application) | For TLC Enantiomer Separation |
| Thickness (Metric) | 0.25 mm |
| Dimensions (L x W) | 10 x 10 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction