missing translation for 'onlineSavingsMsg'
Learn More

Silicon(II) oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis)

Product Code. 15434845
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15434845 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15434845 Supplier Thermo Scientific Alfa Aesar Supplier No. 041093.KS
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10097-28-6 | OSi | 44.084 g/mol

Silicon(II) oxide sputtering target coatings are used for optical applications in reflectors, flood lamps, mirrors, jewelry, and other products. The coatings can reduce reflection in the near infrared range or act as interference layers. It is used in thin-film capacitors, hybrid circuits, semiconductor components and other electronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 10097-28-6
Odor Odorless
Molecular Formula OSi
MDL Number MFCD00151536
Synonym silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
Solubility Information Insoluble in water.
InChI Key LIVNPJMFVYWSIS-UHFFFAOYSA-N
SMILES [O+]#[Si-]
IUPAC Name oxoniumylidynesilanide
Molecular Weight (g/mol) 44.084
PubChem CID 66241
ChEBI CHEBI:30588
Formula Weight 44.09
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Silicon(II) oxide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.