missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) chloride, 99%

Product Code. 11364799
Click to view available options
Quantity:
10 mL
100 mL
1 L
3 x 1 L
Unit Size:
1 L
100mL
10mL
3 L
This item is not returnable. View return policy

Product Code. 11364799

missing translation for 'mfr': Thermo Scientific Alfa Aesar 069182.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-04-7 | Cl4Si | 169.89 g/mol

Silicon(IV) chloride is used as an intermediate in the manufacture of polysilicon. It is also used in the semiconductors industry, photovoltaic cells and in the production of optical fibers. Its derivatives like trichlorosilane and silane are used in the Siemens process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-04-7
Molecular Formula Cl4Si
Molecular Weight (g/mol) 169.89
MDL Number MFCD00011229
InChI Key FDNAPBUWERUEDA-UHFFFAOYSA-N
Synonym silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4
PubChem CID 24816
IUPAC Name tetrachlorosilane
SMILES Cl[Si](Cl)(Cl)Cl

Specifications

Boiling Point 57.6°C
Melting Point °C to 70°C
Color Colorless
Physical Form Liquid
Quantity 100 mL
Assay Percent Range 99% (metals basis)
Solubility Information Miscible with benzene,toluene,chloroform,petroleum ether,carbon tetrachloride,ether and hydrochloric acid.
Formula Weight 169.9
Odor Acrid
Sensitivity Moisture sensitive
Appearance Colourless liquid
Density 1.483
Chemical Name or Material Silicon(IV) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silicon(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Silicon tetrachloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.