missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) nitride, 99.3% (metals basis), typically 90% beta-phase

Product Code. 11323958
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323958 250 g 250g
11313958 10 g 10g
11303958 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323958 Supplier Thermo Scientific Alfa Aesar Supplier No. 040379.30
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 12033-89-5 | N4Si3 | 140.28 g/mol

Silicon(IV) nitride is used in automobile industry as a material for engine parts such as diesel engines and glowplugs. It is used for rocker arm pads for lower wear in spark-ignition engines. Ceramics made from this show good shock resistance. Similarly bearings made from this find application in the main engines of NASA’s space shuttle. It acts as an insulator and in the manufacture of integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12033-89-5
Molecular Formula N4Si3
Molecular Weight (g/mol) 140.28
MDL Number MFCD00011230
InChI Key HQVNEWCFYHHQES-UHFFFAOYSA-N
Synonym silicon nitride, silicon nitride si3n4, unii-qhb8t06idk, qhb8t06idk, silicon iv nitride, alpha phase, trisilicon tetranitride, nierite, nano silicon nitride, silicon iv nitride, silicon nitride, amorphous
PubChem CID 3084099
IUPAC Name Silicon nitride
SMILES N12[Si]34N5[Si]11N3[Si]25N41

Specifications

Melting Point 1900°C (sublimation)
Color Gray to Black
Physical Form Powder
Quantity 250 g
Linear Formula Si3N4
Merck Index 14,8497
Solubility Information Insoluble in water.
Formula Weight 140.28
Percent Purity 99.3%
Odor Odorless
Density 3.2 g/mL
Assay (metals basis)
Chemical Name or Material Silicon(IV) nitride, Typically 90% beta-phase
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.