missing translation for 'onlineSavingsMsg'
Learn More

Silicon oxide, catalyst support, high surface area

Product Code. 11459980
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459980 100 g 100g
11469980 500 g 500g
11479980 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459980 Supplier Thermo Scientific Alfa Aesar Supplier No. 044740.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Form: S.A. 250m2/g | CAS: 7631-86-9 | O2Si | 60.08 g/mol

Typical properties: S.A.250m^2/gSilicon oxide is used as a catalyst support material having a high surface area. It acts as a medium to which catalyst is affixed. Several metals such as platinum, rhodium, ruthenium, silver, copper, nickel, iron, manganese, cobalt, tungsten and gold are supported over silicon dioxide and used as a catalyst in synthetic chemistry for various applications. For example, silica-supported gold catalysts is used for carbon monoxide oxidation by a solution-based technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Spécification

Color White
Physical Form Pellets
Quantity 100 g
Odor Odorless
Appearance White pellets
Solubility Information Insoluble in water.
Chemical Name or Material Silicon oxide
Is Silicon oxide, catalyst support, high surface area sold in powder or pellet form?

Silicon oxide, catalyst support, high surface area comes in pellet form.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.