missing translation for 'onlineSavingsMsg'
Learn More

Silicon powder, -325 mesh, 99.999% (metals basis)

Product Code. 15487275
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487275 250 g 250g
15467275 10 g 10g
15477275 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15487275 Supplier Thermo Scientific Alfa Aesar Supplier No. 046654.30
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-21-3 | Si | 28.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.09
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]

Specifications

Quantity 250 g
Mesh Size −325 Mesh
Odor Odorless
Form Powder
UN Number UN1346
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Silicon powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.