missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Silicone Bottle Holder

Product Code. 15393174 Shop All DWK Life Sciences Products
Click to view available options
Color:
Gray
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15393174

Brand: Duran™ 292135401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Product Type Bottle Holder
Color Gray
Material Silicone
For Use With (Application) Helps to prevent laboratory bottles tipping over when used with pump dispensers or during the filtration
For Use With (Equipment) Round and square bottles
No. per Pack 1 Pack
Quantity 1 Pack
Length (Metric) 75 to 120 mm
Diameter (Metric) Outer 165 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.