missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Silicone Seals

Product Code. 12303389
Change view
Click to view available options
For Use With (Equipment):
GL 18 screw thread
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12303389 GL 18 screw thread Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12303389 Supplier Lenz Laborglasinstrumente™ Supplier No. 01331810
  • ONLINE EXCLUSIVE PRICE
    56.10 €
    33.93 € / Pack of 10
    Save 22.17 €
    39% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Diameter (Metric) Inner 10 mm
Includes PTFE cuff
Material Silicone
Quantity 10/Pk.
Thread Size (Metric) GL18
For Use With (Equipment) GL 18 screw thread
Diameter (Metric) 16 mm
Product Type Seal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.