missing translation for 'onlineSavingsMsg'
Learn More

Lauda™ Silicone Tubes

Brand:  Lauda™ RKJ 016

Additional Details : Weight : 30.00000kg

 View more versions of this product

Product Code. 10179131

  • 42.60 € / 1m
Estimated Shipment: 29-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

For connecting thermostat
Silicone Tubing
1 m
10°C to 100°C
Silicone
8 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers