Learn More
Bollé Safety Silium+ Platinum™ CSP Safety Glasses
Product Code. 15580984 Shop All Bollé Safety Products
Product Code. 15580984 Supplier Bollé Safety Supplier No. SILPCSP
  • ONLINE EXCLUSIVE PRICE
    18.47 €
    12.03 € / Pack of 1
    Save 6.44 €
    34% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Wear a comfortable and ultra lightweight polycarbonate model for applications that require extended wear. The SILIUM+ is an excellent choice, combining style, ergonomics and performances. The permanent PLATINUM coating is applied to both surfaces, making them extremely scratch and fog resistant.
  • Ultra-lightweight for extended wear
  • Ultra lightweight frame
  • Adjustable bridge
  • Non-slip bridge
  • Straight, highly flexible ergonomic temples
  • PLATINUM coating
  • All-round vision
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Mechanical risk - Low energy impact - F; Mechanical risk- High speed particles at extreme temperatures - T; Risks relating to radiations - UV Protection - EN 170 - (2-1.2, 2C-1.2); Solar radiations risks: EN 172
Lens Coating Anti-fog, Scratch-resistant
Frame Color Black
Lens Material Polycarbonate