missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Silodosin
Description
Silodosin is a selective α1A adrenoceptor antagonist (Ki = 0.036 nM). Displays >55 fold selectivity for α1A over α1B and α1D. Inhibits norepinephrine-induced [Ca2+]I increase in α1A expressing CHO cells (IC50 = 0.32 nM). Inhibits norepinephrine-induced contraction of isolated human prostate tissue.
Specifications
Specifications
| Chemical Name or Material | 2,3-Dihydro-1-(3-hydroxypropyl)-5-[(2R)-2-[[2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl]amino]propyl]-1H-indole-7-carboxamide |
| CAS | 160970-54-7 |
| Quantity | 50 mg |
| Molecular Formula | C25H32F3N3O4 |
| Formula Weight | 495.53 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?