Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Product Code. 10661196 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Thermo Scientific DSQ GC-MS Interface
Thermo Scientific ISQ GC-MS Interface
Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Diameter (Metric) Inside:
0.1 to 0.25 mm
0.32 mm
0.53 mm
Unit Size:
Pack of 10
Product Code. 10661196
Brand: Thermo Scientific™ 290MA216
Only null left
Add to basket
Add to basket
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule Kit
Description 290 MA216
For Use With (Equipment) Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Material SilTite Metal
Includes 2 SilTite Nuts, 10 Ferrules, 2 Baseplate Seals
Diameter (Metric) Inside 0.32 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.