Learn More
Thermo Scientific™ SilTite μ-Union Kit for 0.1 to 0.25 mm Column to 0.32 mm Column
Shop All Thermo Scientific ProductsDescription
A high-performance metal union designed for leak-free connections between capillary GC columns of different internal diameters. Its SilTite construction ensures robust sealing, thermal stability, and long-term reliability under GC operating conditions.
- This SilTite μ-Union kit provides a durable, inert metal connection for joining capillary GC columns of varying diameters.
- It delivers secure, leak-free sealing even through repeated high-temperature GC cycles.
- Designed to minimize dead volume and maintain chromatographic performance.
- Its SilTite ferrules form permanent, stable seals without retightening.
- Ideal for column coupling, detector interfacing, or method development requiring flexible GC configurations
Specifications
Specifications
| Type | Valco Union |
| Material | Stainless Steel |
| For Use With (Equipment) | 0.10 to 0.25 mm and 0.32 mm Capillary GC Columns |