missing translation for 'onlineSavingsMsg'
Learn More

Silver, 99.9%, foil, 25x25 mm, 1.0 mm thick, 1ea = 6.6gr

Product Code. 10361324 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
4 Ea.
Unit Size:
Each
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10361324 4 Ea. Pack of 4
10290843 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10361324 Supplier Thermo Scientific Acros Supplier No. 317750040
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Melting Point 961.0°C
Boiling Point 2212.0°C
Quantity 4 Ea.
Assay Percent Range 99.90%
Form Foil
Linear Formula Ag
Fieser 04,164
Merck Index 15, 8643
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hno3, hot h2so4 and alkali cyanide solu, insoluble in alkalies
Formula Weight 107.87
Percent Purity 99.9%
Chemical Name or Material Silver, 99.9%, Each sheet is 25x25mm, 6.6g
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.