missing translation for 'onlineSavingsMsg'
Learn More

Silver, 99.9%, (trace metal basis), wire, 1.0 mm diam., 1ea = 8.2gr

Product Code. 10232512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10232512 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10232512 Supplier Thermo Scientific Acros Supplier No. 317740010
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 7440-22-4 | Ag | 107.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Spezifikation

Color Silver
CAS 7440-22-4
Melting Point 961.0°C
Boiling Point 2212.0°C
Quantity 1 Ea.
Assay Percent Range 99.9%
Form Wire
Molecular Formula Ag
Linear Formula Ag
MDL Number MFCD00003397
Fieser 04,164
Merck Index 15, 8643
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
SMILES [Ag]
IUPAC Name silver
Molecular Weight (g/mol) 107.87
PubChem CID 23954
ChEBI CHEBI:9141
Formula Weight 107.87
Percent Purity 99.9%
Grade Trace Metal Basis
Chemical Name or Material Silver, 99.9% (1ea = 8.2gr)
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.