missing translation for 'onlineSavingsMsg'
Learn More

Silver Acetate, purum p.a, ≥99.0% (T), Honeywell™

Product Code. p-7120758 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15657340

Brand: Honeywell Research Chemicals 8514010G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-63-3
Molecular Formula C2H3AgO2
Molecular Weight (g/mol) 166.912
MDL Number MFCD00012446
InChI Key CQLFBEKRDQMJLZ-UHFFFAOYSA-M
Synonym silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
PubChem CID 11246
IUPAC Name silver;acetate
SMILES CC(=O)[O-].[Ag+]

Specifications

Quantity 10 g
Linear Formula CH3COOAg
UN Number UN3077
Beilstein 3595636
Formula Weight 166.91g/mol
Percent Purity ≥99.0% (T)
Packaging Glass bottle
Chemical Name or Material Silver acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.