missing translation for 'onlineSavingsMsg'
Learn More

Silver Acetate, TRC

Product Code. 30411456
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30411456 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30411456

Brand: Toronto Research Chemicals TRCS4650401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver Acetate
CAS 563-63-3
Molecular Formula C2H3AgO2
Quantity 1 g
Synonym Argentous acetate, Silver Monoacetate
InChI Formula InChI=1S/C2H4O2.Ag/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1
SMILES [Ag+].CC(=O)[O-]
IUPAC Name silver;acetate
Molecular Weight (g/mol) 166.91
Formula Weight 165.9184
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.