Learn More
Silver bromide, Premion™, 99.998% (metals basis)
Description
Silver bromide is used in photographic film. Used in production of radiograph. It also used in infrared applications as windows and lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 432°C |
| Color | Yellow |
| Physical Form | Crystalline |
| Quantity | 5 g |
| UN Number | UN3077 |
| Merck Index | 14,8506 |
| Solubility Information | soluble in alkali cyanide solutions. Partially soluble in ammonia. Insoluble in alcohol and acids. Practically insoluble in water. |
| Formula Weight | 187.78 |
| Percent Purity | 99.998% |
| Odor | Odorless |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.