missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride, 95%

Product Code. 11384969
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384969 25 g 25g
11374969 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384969 Supplier Thermo Scientific Alfa Aesar Supplier No. 087342.14
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silver plating | CAS: 7783-90-6 | AgCl | 143.32 g/mol

Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements. It serves as an antidote for mercury poisoning, and eliminates mercury from body. It is used in glass manufacturing industry. It is useful in the production of bandages, wound healing products and inglaze lustre, personal deodorant products, as well as for long term preservation of drinking water in water tanks; its pharmaceutical composition finds use as an antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1550°C
Melting Point 455°C
Color White
Physical Form Coarse Powder
Quantity 25 g
UN Number UN3077
Merck Index 14,8509
Solubility Information Practically insoluble in water; Soluble in ammonia,solutions of alkali cyanides,thiosulfates and ammonium carbonates; Insoluble in alcohol and dilute acids.
Formula Weight 143.32
Percent Purity 95%
Odor Odorless
Refractive Index 2.071
Sensitivity Light sensitive
Density 5.56 g/mL
Chemical Name or Material Silver chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.