missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride, 99+%, extra pure

Product Code. 10256600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
10g
1kg
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10256600 50 g Glass bottle 50g
10441831 10 g Glass bottle 10g
11944351 250 g Glass bottle 250g
15109243 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10256600 Supplier Thermo Scientific Acros Supplier No. 211270500
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7783-90-6 | AgCl | 143.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1550.0°C
Melting Point 455.0°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99+%
Linear Formula AgCl
Fieser 05,580; 10,347
Merck Index 15,8648
Solubility Information Solubility in water: 1.93 mg/l (25°C). Other solubilities: insoluble in alcohol
Formula Weight 143.32
Percent Purity ≥99%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Silver chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver chloride
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P390-Absorb spillage to prevent material damage.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Silver chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.