missing translation for 'onlineSavingsMsg'
Learn More

Silver Chloride, 99%, ReagentPlus™, Honeywell™

Product Code. 15682260 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15682260 50 g Glass Bottle 50g
15672260 10 g Glass Bottle 10g
15662260 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15682260 Supplier Honeywell Research Chemicals Supplier No. 22792750G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1547°C
Melting Point 455°C
Color White
Quantity 50 g
UN Number UN3077
Formula Weight 143.32g/mol
Percent Purity 99%
Grade ReagentPlus
Packaging Glass Bottle
Appearance Powder or Powder with Chunk(s)
Vapor Pressure 1mmHg (912°C)
Density 5.54g/cm3
Chemical Name or Material Silver Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.