missing translation for 'onlineSavingsMsg'
Learn More

Silver chloride, Purum, ≥99.5%, Honeywell Fluka™

Product Code. p-7111740 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
6 x 100 g
Packaging:
Glass Bottle
Unit Size:
100g
Pack of 6
Product Code. Quantity Packaging unitSize
15630950 100 g Glass Bottle 100g
15640950 6 x 100 g Glass Bottle Pack of 6
2 options
This item is not returnable. View return policy

Product Code. 15630950

Brand: Honeywell-Fluka 10213100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-90-6
Molecular Formula AgCl
Molecular Weight (g/mol) 143.32
MDL Number MFCD00003399
InChI Key HKZLPVFGJNLROG-UHFFFAOYSA-M
IUPAC Name silver(1+) chloride
SMILES [Cl-].[Ag+]

Specifications

Boiling Point 1,547°C
Melting Point 455°C (literature)
CAS Max % 100.0000%
Quantity 100 g
Linear Formula Hn+2PnO3n+1
UN Number UN3260
Formula Weight 143.32
Percent Purity >99.5%
Grade purum
Packaging Glass Bottle
Density 5.54 g/cm3
Chemical Name or Material Silver chloride, Purum, ≥99.5%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.