Learn More
Silver chloride, ultra dry, 99.997% (metals basis)
Beskrivning
It is used in silver plating, electrode, photographic paper, in making antiseptic silver preparations. It is used to make photographic film.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifikationer
Specifikationer
| Boiling Point | 1550°C |
| Melting Point | 455°C |
| Color | White |
| Physical Form | Beads |
| Quantity | 1 g |
| UN Number | UN3077 |
| Merck Index | 14,8509 |
| Solubility Information | Soluble in alkali cyanides,thiosulfates,ammonium carbonates,ammonium chloride,mercuric nitrate,and silver nitrate. Insoluble in water. |
| Formula Weight | 143.32 |
| Percent Purity | 99.997% |
| Visa mer |
RUO – Research Use Only
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.