missing translation for 'onlineSavingsMsg'
Learn More

Silver Diethyldithiocarbamate Silver Salt, Extra Pure, SLR, Fisher Chemical

Product Code. 10346100 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10346100 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10346100 Supplier Fisher Chemical Supplier No. S/1200/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 172°C
CAS 1470-61-7
Color Green
Physical Form Powder/Solid
Quantity 5 g
Molecular Formula C5H10AgNS2
MDL Number 4929
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES CCN(CC)C(=S)[S-].[Ag+]
IUPAC Name silver;N,N-diethylcarbamodithioate
Molecular Weight (g/mol) 256.13
PubChem CID 3034078
Formula Weight 256.13g/mol
Packaging Glass Jar
Show More Show Less
compliance-icons
Product Identifier
  • Silver diethyldithio-carbamate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.