missing translation for 'onlineSavingsMsg'
Learn More

Silver gauze, 80 mesh woven from 0.115mm (0.0045in) dia wire, 99.9% (metals basis)

Product Code. 11355247
Change view
Click to view available options
Quantity:
25 x 25 mm
75 x 75 mm
150 x 150 mm
300 x 300 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11355247 300 x 300 mm Each
11345247 25 x 25 mm Each
11325247 75 x 75 mm Each
11335247 150 x 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11355247 Supplier Thermo Scientific Alfa Aesar Supplier No. 040936.HB
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-22-4 | Ag | 107.87 g/mol

Silver gauze is used for filtration. It is also used in electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-22-4
Molecular Formula Ag
Molecular Weight (g/mol) 107.87
MDL Number MFCD00003397
InChI Key BQCADISMDOOEFD-UHFFFAOYSA-N
Synonym argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
PubChem CID 23954
ChEBI CHEBI:9141
IUPAC Name silver
SMILES [Ag]

Specifications

Color Silver
Appearance Silver-colored guaze
Quantity 300 x 300 mm
Mesh Size 80 Mesh
Odor Characteristic
Assay Percent Range 99.9% (Metals basis)
Form Wire Cloth
Solubility Information Insoluble in water.
Chemical Name or Material Silver

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.