missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluoroantimonate, 98%

Product Code. 10571901 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10571901

Brand: Thermo Scientific Acros 312500050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26042-64-8 | AgF6Sb | 343.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26042-64-8
Molecular Formula AgF6Sb
Molecular Weight (g/mol) 343.61
MDL Number MFCD00003401
InChI Key GSXFODUDOAXUBF-UHFFFAOYSA-H
Synonym silver hexafluoroantimonate, silver hexafluoroantimonate v, silver hexafluorostiboranuide, silver 1+ hexafluorostibanuide, ag.f6sb, silverhexafluoroantimonate, agsbf6, acmc-209tq8, ksc492g5j, silver i hexafluoroantimonate
PubChem CID 16687962
IUPAC Name silver;hexafluoroantimony(1-)
SMILES F[Sb-](F)(F)(F)(F)F.[Ag+]

Specifications

Color White to Yellow
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 97.5% min. (Argentometry)
Linear Formula AgSbF6
Fieser 05,577; 07,320; 17,17
Solubility Information Solubility in water: soluble
Formula Weight 343.61
Percent Purity 98%
Packaging Plastic bottle
Chemical Name or Material Silver hexafluoroantimonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.