missing translation for 'onlineSavingsMsg'
Learn More

Silver hexafluoroarsenate, 98+%, Thermo Scientific Chemicals

Product Code. 11347357
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347357 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11347357 Supplier Thermo Scientific Alfa Aesar Supplier No. 011608.09
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 12005-82-2 | Molecular Formula: AgAsF6 | Molecular Weight: 296.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12005-82-2
Molecular Formula AgAsF6
Molecular Weight (g/mol) 296.78
MDL Number MFCD00014142
InChI Key UNSKNPXMHZPWPB-UHFFFAOYSA-N
Synonym silver hexafluoroarsenate, silver hexafluoroarsenate v, silver hexafluoroarsoranuide, acmc-1c478, silver hexakis fluoranyl arsenic 1-, silver 1+ hexafluoro-??-arsanuide, silver 1+ ion hexafluoro-??-arsanuide
PubChem CID 11077325
IUPAC Name silver;hexafluoroarsenic(1-)
SMILES F[As-](F)(F)(F)(F)F.[Ag+]

Specifications

Color White
Physical Form Powder
Quantity 10 g
Odor Odorless
Sensitivity Moisture sensitive
UN Number UN1557
Formula Weight 296.78
Percent Purity ≥98%
Chemical Name or Material Silver hexafluoroarsenate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.