missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) oxide, 99+%

Product Code. 10751481 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10751481

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20667-12-3 | Ag2O | 231.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Color Black
Physical Form Powder
Quantity 50 g
Assay Percent Range 99+%
Linear Formula Ag2O
Fieser 01,1011; 02,368; 03,252; 04,430; 05,583; 06,515; 07,321; 08,442; 10,350; 11,468
Merck Index 15, 8660
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in dil.hno3 and ammonia,practically insoluble in alcohol
Formula Weight 231.74
Percent Purity ≥99%
Packaging Glass bottle
Chemical Name or Material Silver(I) oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Silver(I) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.