Learn More
Silver(I) oxide, 99.99% (metals basis)
Description
Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 230°C (decomposition) |
| Color | Brown |
| Physical Form | Powder |
| Quantity | 1 g |
| Assay Percent Range | 99.99% (metals basis) |
| Solubility Information | Soluble in dilute nitric acid and ammonia. Practically insoluble in water and alcohol |
| Formula Weight | 231.74 |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Appearance | Dark brown powder |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.