missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
| Color | Black |
| Physical Form | Powder |
| Quantity | 250 g |
| Assay Percent Range | 99+% |
| Linear Formula | Ag2O |
| Fieser | 01,1011; 02,368; 03,252; 04,430; 05,583; 06,515; 07,321; 08,442; 10,350; 11,468 |
| Merck Index | 15, 8660 |
| Solubility Information | Solubility in water: slightly soluble. Other solubilities: freely soluble in dil.hno3 and ammonia,practically insoluble in alcohol |
| Formula Weight | 231.74 |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
Product Identifier
- Silver(I) oxide
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 1
- H271-May cause fire or explosion; strong oxidiser.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Silver oxide
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?