missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) oxide, Electrical Grade

Product Code. 11319378
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319378 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11319378 Supplier Thermo Scientific Alfa Aesar Supplier No. 043268.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For battery plates, pollution control filters | CAS: 20667-12-3 | Ag2O | 231.74 g/mol

For battery plates, and pollution control filters. It is used in the preparation of other silver compounds, and silver-oxide batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20667-12-3
Molecular Formula Ag2O
Molecular Weight (g/mol) 231.74
MDL Number MFCD00003404
InChI Key KHJDQHIZCZTCAE-UHFFFAOYSA-N
IUPAC Name (argentiooxy)silver
SMILES [Ag]O[Ag]

Specifications

Melting Point 230°C (decomposition)
Color Brown
Physical Form Powder
Quantity 250 g
UN Number UN1479
Merck Index 14,8521
Solubility Information Soluble in dilute HNO3 and ammonia; Practically insoluble in water; Insoluble in alcohol
Formula Weight 231.74
Grade Electronic
Odor Odorless
Sensitivity Light sensitive
Density 7.2 g/mL
Chemical Name or Material Silver (I) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.