missing translation for 'onlineSavingsMsg'
Learn More

Silver(I) Perchlorate Anhydrous, TRC

Product Code. 30423318
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30423318 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30423318

Brand: Toronto Research Chemicals TRCS839390100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver(I) Perchlorate Anhydrous
CAS 7783-93-9
Molecular Formula ClO4 . Ag
Quantity 100 mg
Synonym Silver perchlorate, Silver monoperchlorate, Silver perchlorate (AgClO4), Silver(1+) perchlorate, Silver(I) perchlorate
InChI Formula InChI=1S/Ag.ClHO4/c;2-1(3,4)5/h;(H,2,3,4,5)/q+1;/p-1
SMILES [Ag+].[O-]Cl(=O)(=O)=O
IUPAC Name silver;perchlorate
Molecular Weight (g/mol) 207.32
Formula Weight 205.8536
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.