missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) oxide, 85.5-88.0% Ag

Product Code. 10791861 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10791861

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1301-96-8 | AgO | 123.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301-96-8
Molecular Formula AgO
Molecular Weight (g/mol) 123.87
MDL Number MFCD00044285
InChI Key OTCVAHKKMMUFAY-UHFFFAOYSA-N
Synonym silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
PubChem CID 92152
IUPAC Name oxosilver
SMILES O=[Ag]

Specifications

Melting Point >100°C
Color Black to Gray
Physical Form Powder
Quantity 10 g
Linear Formula AgO
Fieser 02,369; 04,431; 06,518; 07,322; 09,412; 10,352; 11,469
Merck Index 15, 8661
Solubility Information Solubility in water: insoluble in water
Formula Weight 123.87
Percent Purity 85.5 to 88% Ag
Packaging Glass bottle
Chemical Name or Material Silver(II) oxide, 85.5-88.0% Ag
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Silver(II) oxide
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.