missing translation for 'onlineSavingsMsg'
Learn More

Silver(II) oxide, 99.9% (metals basis), Ag 86.6% min

Product Code. 11399317
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11399317

Brand: Thermo Scientific Alfa Aesar 040187.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1301-96-8 | AgO | 123.867 g/mol

Silver(II) oxide is used to make silver oxide-zinc alkali batteries. It is also used as a reagent in laboratory reactions and forms transition metal-carbene complexes. It is also used as a catalyst to prepare silver metal. It acts as an effective carbon dioxide scrubber and used in nuclear submarines, the international space station and space shuttles. Further, it is used in the manufacture of filters for gas sensors and in some infection-resistant surgical fabric materials and fibrous textile articles. In addition to this, it is used in concrete and in some swimming pools to protect the water from undesired microbes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1301-96-8
Molecular Formula AgO
Molecular Weight (g/mol) 123.867
MDL Number MFCD00044285
InChI Key OTCVAHKKMMUFAY-UHFFFAOYSA-N
Synonym silver ii oxide, silver oxide, silberperoxyd, silver oxide,ago, acmc-1aiyc, silver i,iii oxide, ksc203c6r, silver ii oxide 5g, silver ii oxide-ag
PubChem CID 92152
IUPAC Name oxosilver
SMILES O=[Ag]

Specifications

Melting Point >100°C (decomposition)
Color Black to Gray
Physical Form Powder
Quantity 5 g
UN Number UN3085
Merck Index 14,8522
Solubility Information Soluble in alkalis. Insoluble in water.
Formula Weight 123.87
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Ag 86.6% min.
Odor Odorless
Sensitivity Light and moisture sensitive
Density 7.44 g/mL
Assay (metals basis)
Chemical Name or Material Silver(II) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.