missing translation for 'onlineSavingsMsg'
Learn More

Silver iodide, Premion™, 99.999% (metals basis)

Product Code. 11396468
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11396468

Brand: Thermo Scientific Alfa Aesar 012111.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-96-2 | AgI | 234.773 g/mol

Silver iodide is used in photographic film. Used in medical devices as a sustained release antibacterial. It also used in as an antiseptic and in cloud seeding. It is used in proteomics research applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Photographic developers, generating SO{2}, reducing agent (dyes), textile bleaching (antichlor)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-96-2
Molecular Formula AgI
Molecular Weight (g/mol) 234.773
MDL Number MFCD00003412
InChI Key MSFPLIAKTHOCQP-UHFFFAOYSA-M
Synonym silver iodide, neosiluol, neosilvol, silver monoiodide, silver 1+ iodide, colloidal silver iodide, iodanylsilver, silver iodide metals basis, ccris 8490
PubChem CID 24563
IUPAC Name iodosilver
SMILES [Ag]I

Specifications

Melting Point 558°C
Color Yellow
Physical Form Powder
Quantity 100 g
UN Number UN3077
Merck Index 14,8516
Solubility Information Soluble in alkali cyanides and iodides. Insoluble in water,acids and ammonium carbonate.
Formula Weight 234.77
Percent Purity 99.999%
Grade Premion
Odor Odorless
Sensitivity Light sensitive
Density 5.67 g/mL
Assay (metals basis)
Chemical Name or Material Silver iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.